Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053812.1 | NP_446264.1 | 2 - 3 | 315 | 64 | |
XM_006256101.2 | XP_006256163.1 | 2 - 3 | 334 | 64 | ||
GenBank mRNA | AF259504.1 | - | 2 - 3 | 315 | 64 | |
BC089784.1 | - | 2 - 3 | 276 | 7366192 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |