Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053844.1 | NP_446296.1 | 2 - 3 | 258 | 110 | |
GenBank mRNA | M97255.1 | - | 2 - 3 | 258 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |