Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057120.1 | NP_476461.1 | 2 - 3 | 311 | 78 | |
XM_006248911.2 | XP_006248973.1 | 3 - 4 | 362 | 78 | ||
GenBank mRNA | D49977.1 | - | 2 - 3 | 311 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |