Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057132.3 | NP_476473.1 | 3 - 4 | 407 | 62 | |
XM_006243699.1 | XP_006243761.1 | 5 - 6 | 675 | 62 | ||
XM_006243700.1 | XP_006243762.1 | 5 - 6 | 650 | 62 | ||
XM_006243701.1 | XP_006243763.1 | 5 - 6 | 770 | 62 | ||
GenBank mRNA | AY026068.1 | - | 3 - 4 | 407 | 62 | |
AY026069.1 | - | 3 - 4 | 407 | 62 | ||
BC061732.1 | - | 5 - 6 | 584 | 5598621 | 62 | |
D84477.1 | - | 3 - 4 | 377 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |