Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057151.1 | NP_476492.1 | 1 - 2 | 70 | 73 | |
GenBank mRNA | AF312687.1 | - | 1 - 2 | 70 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Rat Chemokine Panel Inventoried Probe spans exons |