Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301285.1 | NP_001288214.1 | 5 - 6 | 649 | 88 | |
NM_001301286.1 | NP_001288215.1 | 4 - 5 | 473 | 88 | ||
NM_057208.2 | NP_476556.2 | 4 - 5 | 473 | 88 | ||
XM_006232572.2 | XP_006232634.1 | 5 - 6 | 669 | 88 | ||
XM_006232573.2 | XP_006232635.1 | 5 - 6 | 669 | 88 | ||
XM_006232575.2 | XP_006232637.1 | 5 - 6 | 669 | 88 | ||
XM_006232577.2 | XP_006232639.1 | 5 - 6 | 669 | 88 | ||
XM_006232581.2 | XP_006232643.1 | 4 - 5 | 574 | 88 | ||
XM_006232583.2 | XP_006232645.1 | 4 - 5 | 577 | 88 | ||
GenBank mRNA | FQ214881.1 | - | 5 - 6 | 647 | 88 | |
FQ215580.1 | - | 5 - 6 | 649 | 88 | ||
FQ215752.1 | - | 5 - 6 | 649 | 88 | ||
FQ216429.1 | - | 5 - 6 | 648 | 88 | ||
L24776.1 | - | 473 | 88 | |||
L24777.1 | - | 4 - 5 | 473 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus Inventoried Probe spans exons |