Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133297.2 | NP_579831.2 | 3 - 4 | 580 | 85 | |
GenBank mRNA | AF390544.1 | - | 3 - 4 | 304 | 85 | |
BC060547.1 | - | 3 - 4 | 329 | 6921948 | 85 | |
CK596377.1 | - | 3 - 4 | 595 | 85 | ||
DY561321.1 | - | 3 - 4 | 318 | 85 | ||
FQ209574.1 | - | 3 - 4 | 315 | 85 | ||
FQ212727.1 | - | 3 - 4 | 348 | 85 | ||
FQ213177.1 | - | 3 - 4 | 329 | 85 | ||
FQ213838.1 | - | 3 - 4 | 315 | 85 | ||
FQ214634.1 | - | 3 - 4 | 325 | 85 | ||
FQ215362.1 | - | 3 - 4 | 315 | 85 | ||
FQ215992.1 | - | 3 - 4 | 338 | 85 | ||
FQ216831.1 | - | 3 - 4 | 315 | 85 | ||
FQ219237.1 | - | 3 - 4 | 336 | 85 | ||
FQ219570.1 | - | 3 - 4 | 315 | 85 | ||
FQ219725.1 | - | 3 - 4 | 308 | 85 | ||
FQ220311.1 | - | 3 - 4 | 314 | 85 | ||
FQ220593.1 | - | 3 - 4 | 320 | 85 | ||
FQ229133.1 | - | 3 - 4 | 321 | 85 | ||
FQ230131.1 | - | 3 - 4 | 315 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |