Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133615.1 | NP_598299.1 | 5 - 6 | 475 | 79 | |
XM_006256993.2 | XP_006257055.1 | 6 - 7 | 942 | 79 | ||
XM_006256994.2 | XP_006257056.1 | 6 - 7 | 941 | 79 | ||
GenBank mRNA | BC090332.1 | - | 5 - 6 | 530 | 7193160 | 79 |
U40188.1 | - | 5 - 6 | 475 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |