Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134384.1 | NP_599211.1 | 4 - 5 | 348 | 71 | |
GenBank mRNA | AB057408.1 | - | 4 - 5 | 348 | 71 | |
BC081729.1 | - | 4 - 5 | 365 | 7132055 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |