Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134409.2 | NP_599236.2 | 3 - 4 | 217 | 75 | |
GenBank mRNA | AB095177.1 | - | 3 - 4 | 217 | 75 | |
CF249177.1 | - | 2 - 3 | 189 | 75 | ||
Z30584.1 | - | 3 - 4 | 246 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |