Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134468.1 | NP_604463.1 | 10 - 11 | 1025 | 69 | |
XM_006236999.2 | XP_006237061.1 | 11 - 12 | 1694 | 69 | ||
XM_006237000.2 | XP_006237062.1 | 11 - 12 | 1690 | 69 | ||
XM_006237001.2 | XP_006237063.1 | 10 - 11 | 1557 | 69 | ||
XM_006237002.2 | XP_006237064.1 | 10 - 11 | 1484 | 69 | ||
XM_006237003.2 | XP_006237065.1 | 10 - 11 | 1483 | 69 | ||
GenBank mRNA | BC071177.1 | - | 10 - 11 | 960 | 7098592 | 69 |
FQ213485.1 | - | 988 | 69 | |||
L26288.1 | - | 10 - 11 | 1025 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |