Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138505.2 | NP_612514.2 | 1 | 1451 | 63 | |
GenBank mRNA | M32061.1 | - | 1 | 1448 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |