Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139116.1 | NP_620816.1 | 3 - 4 | 217 | 71 | |
XM_006238021.2 | XP_006238083.1 | 3 - 4 | 845 | 71 | ||
XM_006238022.2 | XP_006238084.1 | 3 - 4 | 265 | 71 | ||
XM_006238023.2 | XP_006238085.1 | 3 - 4 | 837 | 71 | ||
GenBank mRNA | AF347936.1 | - | 3 - 4 | 217 | 71 | |
BC070921.1 | - | 3 - 4 | 212 | 7105297 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |