Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144730.1 | NP_653331.1 | 1 - 2 | 78 | 56 | |
GenBank mRNA | L22761.1 | - | 1 - 2 | 78 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |