Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153722.2 | NP_714944.1 | 1 - 2 | 156 | 86 | |
GenBank mRNA | M35297.1 | - | 159 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Inventoried Probe spans exons |