Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172029.2 | NP_742026.1 | 4 - 5 | 414 | 62 | |
GenBank mRNA | BC081709.1 | - | 4 - 5 | 414 | 7135576 | 62 |
FQ217438.1 | - | 4 - 5 | 452 | 62 | ||
U05821.1 | - | 4 - 5 | 433 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Endogenous Controls Recommended Assay Rattus norvegicus Rat Endogenous Controls 1700 Inventoried Rat_Endogenous_Controls Probe spans exons |