Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcnkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173103.1 | NP_775126.1 | 16 - 17 | 1870 | 71 | |
XM_006239320.2 | XP_006239382.1 | 19 - 20 | 4742 | 71 | ||
XM_008764293.1 | XP_008762515.1 | 18 - 19 | 4410 | 71 | ||
XM_008764294.1 | XP_008762516.1 | 18 - 19 | 2676 | 71 | ||
XM_008764295.1 | XP_008762517.1 | 17 - 18 | 2821 | 71 | ||
XM_008764296.1 | XP_008762518.1 | 16 - 17 | 2699 | 71 | ||
XM_008764297.1 | XP_008762519.1 | 16 - 17 | 2681 | 71 | ||
GenBank mRNA | D26111.1 | - | 17 - 18 | 1924 | 71 | |
DQ437671.1 | - | 17 - 18 | 1974 | 71 | ||
DQ437672.1 | - | 17 - 18 | 1867 | 71 | ||
DQ437673.1 | - | 16 - 17 | 1845 | 71 | ||
DQ437674.1 | - | 16 - 17 | 1809 | 71 | ||
DQ437675.1 | - | 17 - 18 | 1817 | 71 | ||
DQ437676.1 | - | 16 - 17 | 1795 | 71 | ||
Z30663.1 | - | 16 - 17 | 1870 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |