Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173294.2 | NP_775416.1 | 4 - 5 | 677 | 96 | |
GenBank mRNA | BC088103.1 | - | 4 - 5 | 642 | 96 | |
BC105823.1 | - | 4 - 5 | 642 | 96 | ||
BC127479.1 | - | 4 - 5 | 658 | 7372256 | 96 | |
M20406.1 | - | 4 - 5 | 677 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |