Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108180.1 | NP_001101650.1 | 1 - 2 | 533 | 56 | |
XM_006243657.2 | XP_006243719.1 | 1 - 2 | 533 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |