Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC166821.1 | - | 1219 | 8368184 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |