Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012559.2 | NP_036691.2 | 4 - 5 | 424 | 88 | |
XM_006232525.1 | XP_006232587.1 | 4 - 5 | 578 | 88 | ||
GenBank mRNA | BC078893.1 | - | 4 - 5 | 424 | 7122366 | 88 |
FQ218666.1 | - | 2 - 3 | 226 | 88 | ||
FQ219408.1 | - | 4 - 5 | 444 | 88 | ||
FQ219511.1 | - | 4 - 5 | 446 | 88 | ||
FQ219604.1 | - | 4 - 5 | 445 | 88 | ||
FQ229132.1 | - | 4 - 5 | 451 | 88 | ||
J00734.1 | - | 4 - 5 | 409 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |