Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012626.2 | NP_036758.1 | 3 - 4 | 358 | 84 | |
GenBank mRNA | BP484139.1 | - | 3 - 4 | 341 | 84 | |
M13588.1 | - | 3 - 4 | 358 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |