Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012753.2 | NP_036885.1 | 1 - 2 | 331 | 81 | |
XM_006231434.2 | XP_006231496.1 | 2 - 3 | 654 | 81 | ||
XM_006231435.2 | XP_006231497.1 | 2 - 3 | 623 | 81 | ||
GenBank mRNA | BC078898.1 | - | 1 - 2 | 331 | 7111017 | 81 |
M22204.1 | - | 1 - 2 | 333 | 81 | ||
M31681.1 | - | 1 - 2 | 343 | 81 | ||
X14086.1 | - | 1 - 2 | 333 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |