Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021576.2 | NP_067587.2 | 6 - 7 | 1252 | 97 | |
GenBank mRNA | BC081806.1 | - | 6 - 7 | 1282 | 7134340 | 97 |
BC098665.1 | - | 6 - 7 | 1252 | 7322840 | 97 | |
J05214.1 | - | 6 - 7 | 1602 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |