Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021595.2 | NP_067606.1 | 3 - 4 | 577 | 105 | |
XM_008763255.1 | XP_008761477.1 | 3 - 4 | 1087 | 105 | ||
XM_008763256.1 | XP_008761478.1 | 4 - 5 | 1004 | 105 | ||
XM_008763257.1 | XP_008761479.1 | 4 - 5 | 763 | 105 | ||
XM_008763258.1 | XP_008761480.1 | 4 - 5 | 914 | 105 | ||
XM_008763259.1 | XP_008761481.1 | 3 - 4 | 723 | 105 | ||
GenBank mRNA | AB040815.1 | - | 3 - 4 | 578 | 105 | |
AF250322.1 | - | 3 - 4 | 578 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Probe spans exons |