Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022388.1 | NP_071783.1 | 5 - 6 | 1098 | 57 | |
XM_006237206.2 | XP_006237268.1 | 6 - 7 | 437 | 57 | ||
XM_008763260.1 | XP_008761482.1 | 6 - 7 | 767 | 57 | ||
XM_008763261.1 | XP_008761483.1 | 6 - 7 | 725 | 57 | ||
GenBank mRNA | L41254.1 | - | 5 - 6 | 1098 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |