Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022860.2 | NP_074051.1 | 10 - 11 | 1523 | 76 | |
XM_006241497.2 | XP_006241559.1 | 10 - 11 | 2050 | 76 | ||
XM_008765419.1 | XP_008763641.1 | 10 - 11 | 3879 | 76 | ||
XM_008765420.1 | XP_008763642.1 | 9 - 10 | 2200 | 76 | ||
GenBank mRNA | BC081799.1 | - | 10 - 11 | 1523 | 7133837 | 76 |
D17809.1 | - | 10 - 11 | 1412 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |