Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024143.2 | NP_077057.1 | 6 - 7 | 1516 | 88 | |
GenBank mRNA | AF242189.1 | - | 6 - 7 | 1517 | 88 | |
BC091147.1 | - | 6 - 7 | 1539 | 7370589 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |