Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024160.1 | NP_077074.1 | 4 - 5 | 330 | 81 | |
GenBank mRNA | AJ295951.1 | - | 4 - 5 | 316 | 81 | |
U18729.1 | - | 4 - 5 | 330 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |