Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024371.2 | NP_077347.1 | 9 - 10 | 1231 | 88 | |
XM_006237059.1 | XP_006237121.1 | 10 - 11 | 1440 | 88 | ||
XM_006237060.2 | XP_006237122.1 | 10 - 11 | 3660 | 88 | ||
GenBank mRNA | M59742.1 | - | 9 - 10 | 1231 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |