Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031040.1 | NP_112302.1 | 2 - 3 | 1134 | 84 | |
XM_008763199.1 | XP_008761421.1 | 2 - 3 | 1134 | 84 | ||
GenBank mRNA | D16817.1 | - | 2 - 3 | 1134 | 84 | |
U06832.1 | - | 2 - 3 | 869 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |