Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031153.2 | NP_112415.1 | 8 - 9 | 1505 | 71 | |
GenBank mRNA | AF203906.1 | - | 8 - 9 | 1179 | 71 | |
BC083553.1 | - | 8 - 9 | 1505 | 7189111 | 71 | |
FQ213170.1 | - | 8 - 9 | 1123 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |