Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031595.1 | NP_113783.1 | 2 - 3 | 389 | 56 | |
GenBank mRNA | BC062019.1 | - | 2 - 3 | 299 | 5620971 | 56 |
D83522.1 | - | 2 - 3 | 263 | 56 | ||
FQ225567.1 | - | 1 - 2 | 48 | 56 | ||
FQ225947.1 | - | 1 - 2 | 48 | 56 | ||
FQ226273.1 | - | 1 - 2 | 48 | 56 | ||
FQ232987.1 | - | 1 - 2 | 48 | 56 | ||
U77918.1 | - | 2 - 3 | 389 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |