Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031613.1 | NP_113801.1 | 4 - 5 | 557 | 104 | |
XM_006243407.2 | XP_006243469.1 | 5 - 6 | 1017 | 104 | ||
XM_006243408.2 | XP_006243470.1 | 5 - 6 | 4219 | 104 | ||
XM_008766331.1 | XP_008764553.1 | 5 - 6 | 609 | 104 | ||
GenBank mRNA | U59240.1 | - | 4 - 5 | 557 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |