Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031719.1 | NP_113907.1 | 2 - 3 | 297 | 95 | |
XM_006229792.2 | XP_006229854.1 | 2 - 3 | 360 | 95 | ||
GenBank mRNA | BC060555.1 | - | 2 - 3 | 341 | 6918578 | 95 |
D13985.1 | - | 2 - 3 | 367 | 95 | ||
D17698.1 | - | 2 - 3 | 280 | 95 | ||
L26450.1 | - | 2 - 3 | 297 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |