Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031811.3 | NP_113999.2 | 6 - 7 | 847 | 67 | |
GenBank mRNA | AF069306.1 | - | 6 - 7 | 833 | 67 | |
BC059126.1 | - | 6 - 7 | 851 | 6889440 | 67 | |
FQ219168.1 | - | 6 - 7 | 873 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |