Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032063.2 | NP_114452.1 | 2 - 3 | 638 | 71 | |
GenBank mRNA | U78889.2 | - | 2 - 3 | 638 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |