Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053463.1 | NP_445915.1 | 2 - 3 | 241 | 82 | |
XM_006229163.1 | XP_006229225.1 | 3 - 4 | 358 | 82 | ||
XM_006229164.2 | XP_006229226.1 | 3 - 4 | 302 | 82 | ||
XM_006229165.1 | XP_006229227.1 | 3 - 4 | 358 | 82 | ||
GenBank mRNA | AF336828.1 | - | 2 - 3 | 241 | 82 | |
BC062084.1 | - | 3 - 4 | 302 | 82 | ||
BC100643.1 | - | 3 - 4 | 291 | 7930904 | 82 | |
Z36277.1 | - | 3 - 4 | 303 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |