Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053521.1 | NP_445973.1 | 7 - 8 | 1124 | 99 | |
XM_006244251.2 | XP_006244313.1 | 7 - 8 | 1227 | 99 | ||
XM_006244253.2 | XP_006244315.1 | 5 - 6 | 771 | 99 | ||
XM_008766774.1 | XP_008764996.1 | 7 - 8 | 1275 | 99 | ||
XM_008766775.1 | XP_008764997.1 | 7 - 8 | 1224 | 99 | ||
GenBank mRNA | AB030947.1 | - | 7 - 8 | 1124 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |