Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053674.1 | NP_446126.1 | 1 - 2 | 80 | 79 | |
GenBank mRNA | AF121345.1 | - | 1 - 2 | 80 | 79 | |
BC086573.1 | - | 1 - 2 | 75 | 7306754 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |