Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053674.1 | NP_446126.1 | 6 - 7 | 688 | 67 | |
GenBank mRNA | AF121345.1 | - | 6 - 7 | 688 | 67 | |
BC086573.1 | - | 6 - 7 | 683 | 7306754 | 67 | |
FQ216943.1 | - | 4 - 5 | 520 | 67 | ||
FQ224924.1 | - | 4 - 5 | 520 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |