Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053777.1 | NP_446229.1 | 6 - 7 | 1675 | 71 | |
GenBank mRNA | AF092450.1 | - | 5 - 6 | 1410 | 71 | |
AF108959.1 | - | 5 - 6 | 1613 | 71 | ||
AF109772.1 | - | 5 - 6 | 1522 | 71 | ||
AF109773.1 | - | 5 - 6 | 1585 | 71 | ||
AF109774.1 | - | 6 - 7 | 1675 | 71 | ||
DQ377223.1 | - | 5 - 6 | 1485 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |