Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053911.2 | NP_446363.1 | 3 - 4 | 422 | 76 | |
XM_006228964.2 | XP_006229026.1 | 3 - 4 | 440 | 76 | ||
XM_008759304.1 | XP_008757526.1 | 2 - 3 | 913 | 76 | ||
GenBank mRNA | FQ229744.1 | - | 2 - 3 | 143 | 76 | |
U83896.1 | - | 3 - 4 | 426 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |