Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053965.2 | NP_446417.2 | 6 - 7 | 706 | 93 | |
GenBank mRNA | BC081749.1 | - | 6 - 7 | 706 | 7134670 | 93 |
FQ209681.1 | - | 6 - 7 | 662 | 93 | ||
X97831.1 | - | 6 - 7 | 663 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |