Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057198.2 | NP_476546.1 | 7 - 8 | 1443 | 108 | |
GenBank mRNA | BC086999.1 | - | 7 - 8 | 1443 | 7189259 | 108 |
D10853.1 | - | 7 - 8 | 951 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |