Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057198.2 | NP_476546.1 | 9 - 10 | 1798 | 108 | |
GenBank mRNA | BC086999.1 | - | 9 - 10 | 1798 | 7189259 | 108 |
D10853.1 | - | 9 - 10 | 1306 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |