Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057205.2 | NP_476553.1 | 4 - 5 | 267 | 149 | |
XM_006236936.2 | XP_006236998.1 | 3 - 4 | 250 | 149 | ||
GenBank mRNA | AF336829.1 | - | 4 - 5 | 276 | 149 | |
BC081743.1 | - | 4 - 5 | 267 | 7124631 | 149 | |
FQ234407.1 | - | 3 - 4 | 226 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |