Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Des | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022531.1 | NP_071976.1 | 5 - 6 | 1099 | 64 | |
GenBank mRNA | BC061872.1 | - | 5 - 6 | 1092 | 5598951 | 64 |
FQ216012.1 | - | 1 - 2 | 65 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |