Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022629.1 | NP_072151.1 | 5 - 6 | 833 | 87 | |
GenBank mRNA | AF160958.1 | - | 5 - 6 | 833 | 87 | |
FQ210746.1 | - | 925 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |