Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022668.2 | NP_073159.2 | 6 - 7 | 796 | 103 | |
GenBank mRNA | BC087717.1 | - | 6 - 7 | 796 | 7317044 | 103 |
M99485.1 | - | 6 - 7 | 796 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |